Fisherbrand™ Powder Funnels Edge
Edge
Parallel stems minimize bridging of powder.
Supplier:  Fisherbrand™ 3007000100
 View more versions of this product
Catalog No. 10-500-2
Edge
Explore available promotions
Description
Description
Parallel stems minimize bridging of powder
  • Durable polypropylene
  • External ribbing prevents air locks
  • Autoclavable
Specifications
Specifications
10.5cm
21mm
10.3 cm
33mm
225 mL
Polypropylene
Autoclavable