Fisherbrand™ PrepSep™ Large-Volume SPE Columns
Versatile design provides rapid flowthrough using syringe, vacuum box, or robot technology
$1877.25 - $1877.25
Specifications
Column Type SPE
View More Specs
Products 1
Catalog Number Mfr. No. Column Capacity (Metric) Sorbent Price Quantity  
Catalog Number Mfr. No. Column Capacity (Metric) Sorbent Price Quantity  
P452
View Documents
Fisherbrand™
P452
12 mL C8
Pack of 50 Each for $1,877.25
Description
Description
  • Sorbent packings separate and retain sample analytes from their matrices; retrieve analytes by eluting with an appropriate solvent
  • Packing type is clearly marked on upper rim of each column for easy identification
  • Economically disposable; inexpensive enough to use once then throw away, eliminating cross-contamination and cleanups
  • Large-reservoir columns feature 12 to 60mL reservoirs that accept larger sample volumes; also feature a conical shape for easy filling
Specifications
Specifications
SPE
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'