Fisherbrand™ Pressure and Vacuum Rubber Tubing
For pressure and vacuum work
 View more versions of this product
Catalog No. 14-173C
Explore available promotions
Description
Description
For pressure and vacuum work
  • Made with dry natural rubber
  • Available in variety of sizes
  • Offered in Black and Red
Specifications
Specifications
Pressure and Vacuum Tubing
6.4 mm
5/8 in.
12 ft.
0.187 in.
Pressure and vacuum work
1/4 in.
Rubber
15.9 mm
3.7 m
4.8 mm
Red