Fisherbrand™ Pressure and Vacuum Rubber Tubing
For pressure and vacuum work
 View more versions of this product
Catalog No. 14-173D
Explore available promotions
Description
Description
For pressure and vacuum work
  • Made with dry natural rubber
  • Available in variety of sizes
  • Offered in Black and Red
Specifications
Specifications
Pressure and Vacuum Tubing
9.5 mm
1 in.
12 ft.
0.312 in.
Pressure and vacuum work
3/8 in.
Rubber
25.4 mm
3.7 m
7.9 mm
Red