Fisherbrand™ Pressure and Vacuum Rubber Tubing
For pressure and vacuum work
 View more versions of this product
Catalog No. 14-175B
Explore available promotions
Description
Description
For pressure and vacuum work
  • Made with dry natural rubber
  • Available in variety of sizes
  • Offered in Black and Red
Specifications
Specifications
Pressure and Vacuum Tubing
4.8 mm
9/16 in.
12 ft.
0.187 in.
Pressure and vacuum work
0.19 in.
Rubber
14.3 mm
3.7 m
4.8 mm
Black