Fisherbrand™ Pressure and Vacuum Rubber Tubing
For pressure and vacuum work
 View more versions of this product
Catalog No. 14-175E
Explore available promotions
Description
Description
For pressure and vacuum work
  • Made with dry natural rubber
  • Available in variety of sizes
  • Offered in Black and Red
Specifications
Specifications
Pressure and Vacuum Tubing
9.5 mm
7/8 in.
12 ft.
0.25 in.
Pressure and vacuum work
3/8 in.
Rubber
22.2 mm
3.7 m
6.4 mm
Black