missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ PTFE Disposable Stir Bars

The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.

 View more versions of this product

Catalog No. 14-513-93


Explore available promotions

Description

Description

The multifaceted magnetic stirring bar surface adds turbulence relative to similar size smooth cylindrical stirring bars.

  • Ideal for single use applications mixing radioactive reagents or when cross contamination might be a concern
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable

Specifications

Specifications

3.2 mm
PTFE
Stir Bar
100
Autoclavable
12.7 mm
0.5 in.
0.125 in.
White
PTFE
ISO 9001:2008
Cylinder
SDS
Documents

Documents

Accessories
Promotions

Promotions