missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ 3-Place Vacuum Manifold, PVC

For testing multiple samples by vacuum filtration

Supplier:  Fisherbrand™ SC1M033V00

Catalog No. 09-753-39A


Explore available promotions

Description

Description

  • Accommodate vacuum filter holders and funnels using a No. 8 stopper
  • Lightweight design efficiently uses lab space
  • Each holder has two-way valve made of Type 316 stainless-steel with Viton™ seals
  • Manifold body and funnel holders made of polyvinylchloride
  • 1/4 NPTM connections to vacuum
  • Not autoclavable; sanitize using ethanol or formaldehyde
  • Maximum operating temperature: 55°C (130°F)

Specifications

Specifications

3-Place
Vacuum Manifold
4.75 in.
PVC
17.75 in.
6.75 in.
Videos
SDS
Documents

Documents

Promotions

Promotions