Fisherbrand™ Polypropylene, Spigot
For Use With: Fisherbrand HDPE and Polypropylene Rectangular Carboys
Catalog No. 03-007-650
Explore available promotions
Description
Description
Value your lab can rely on
Spigots for use with Fisherbrand HDPE and Polypropylene rectangular carboys
  • Spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing
  • Spigot spout without the adapter fits 5/8-in. ID tubing
Specifications
Specifications
Autoclavable
Polypropylene
Spigot
Fisherbrand HDPE and Polypropylene Rectangular Carboys
1 Ea.