Fisherbrand™ Polypropylene, Closure
For Use With: Fisherbrand HDPE and Polypropylene Rectangular Carboys
Catalog No. 03-007-649
Explore available promotions
Description
Description
Value your lab can rely on
Closures for use with Fisherbrand HDPE and Polypropylene rectangular carboys
  • Closure for the Polypropylene carboys for use when autoclaving with liquid contained inside
Specifications
Specifications
Autoclavable
Fisherbrand HDPE and Polypropylene Rectangular Carboys
Polypropylene
Closure