Fisherbrand™ Redi-Stor™ Conductivity Probe Storage Solution
Ideal for storing conductivity probes.
Description
Ideal for storing conductivity probes.
- Preserves probe's cleanliness
- Eliminates growths found when probes are stored in water
- Keeps probes ready for immediate use with no conditioning
Order Info
16 oz. (473mL) bottle
Specifications
Storage Solution | |
16 oz. | |
Conductivity probe |
Storing conductivity probes | |
454 mL |