Fisherbrand™ Replacement Hydrometers for FBA11582 Precision Specific Gravity Hydrometer Set
Replacement Hydrometers for the FBA11582 Hydrometer Set.
Description
Replacement Hydrometers for the FBA11582 Hydrometer Set.
Replacement Hydrometer for FBA11582 Hydrometer Set.Specifications
0.002 | |
300 mm | |
Soda Lime Glass | |
1.600 to 1.820 |
Without 3-pt Certification | |
FBA11582 Hydrometer Set | |
Specific Gravity Measurement |