Fisherbrand™ Reusable Glass Berzelius Beakers
Made with Borosilicate glass
 View more versions of this product
Catalog No. FB102-1000
Explore available promotions
Description
Description
Fisherbrand™ Reusable Glass Berzelius Beakers feature a convenient tall-form design. Made with Borosilicate glass.
  • Slighty flared top, reinforcing bead and pouring spout
  • Durable matte finish marking area
  • White scales to approximate volumes
  • Made with Borosilicate glass
Compliance
  • Designed from ASTM™ Specification E960, Type III requirements
Specifications
Specifications
33.81 oz.
9 cm
Borosilicate Glass
50 to 950 mL
18.8 cm
50 mL
6 Pk.
Reusable
ASTM E960
Berzelius
3.54 in.
1000 mL
Offer Excellent Mechanical Strength and Durability, While Providing High Resistance to Chemical Attack and Thermal Shock, Ideal for Use When Performing Titrations
7.4 in.
Spout
Matte Finish Marking
Beaker
Yes
Dual Scale
Videos
SDS
Documents
Documents
Accessories