Fisherbrand™ Reusable Glass Long-Stem Funnels
Promote faster, more effective filtering
 View more versions of this product
Catalog No. FB6005865
Explore available promotions
Description
Description
  • Constructed with heavy uniform walls, molded rims and fire-polished stems
  • Formed to a 58° angle for rapid filtration
  • Cone is suspended by its uppermost edge, leaving most of the conical area for filtration
  • High resistance to chemical attack, mechanical and thermal shock
Compliance
  • Designed from ASTM™ Specifications E 1095, Type I, Class B requirements
Specifications
Specifications
6.5cm
8.27 in.
210 mm
58°
11cm
15cm
60 mL
Borosilicate Glass
ASTM E1095, Type I, Class B
65mm
Provides faster, more effective filtering
Videos
SDS
Documents
Documents
Accessories