Fisherbrand™ Reusable Glass Short-Stem Funnels
High resistance to chemical attack, mechanical, and thermal shock
Description
High resistance to chemical attack, mechanical, and thermal shock
- Constructed with heavy uniform walls, molded rims, and fire-polished stems
- Formed to a 58° angle for faster, more effective filtering
- Cone is suspended by its uppermost edge, leaving most of the conical area for filtration
Compliance
- Designed from ASTM™ Specification E 1095, Type I, Class A requirements
Specifications
75mm | |
8mm | |
ASTM E1095, Type I, Class A | |
125mm |
90 mL | |
Borosilicate Glass | |
58° | |
75mm |