Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks
High resistance to heat, chemical attack, and mechanical shock
 View more versions of this product
Catalog No. FB5012000
Explore available promotions
Description
Description
Fisherbrand™ Reusable Glass Wide-Mouth Erlenmeyer Flasks offer sturdy borosilicate glass construction with a heavy-duty, tooled-top finish. High resistance to heat, chemical attack, and mechanical shock.
  • Capacity scale measures approximate volume
  • Matte-finish ceramic marking spot takes writing and erases easily
Specifications
Specifications
Top style: Heavy duty tooled with capacity scale
2000 mL
600 to 2000 mL
10.4 in.
1
Reusable
160 mm
6.33 in.
Borosilicate Glass
100 mL
26.5 cm
Matte Finish Ceramic Enamel Marking
Wide
13