Fisherbrand™ Ringed Cytology Slides
Designed to collect a thin layer of cells in the ringed area
Supplier:  Fisherbrand™ ES333W72600621
 View more versions of this product
Catalog No. 22-037-240
Explore available promotions
Description
Description
  • Fully compatible with Fisherbrand and Epredia™ Shandon™ Cytology Funnels
  • Available with single or double rings, with or without adhesive coating
Specifications
Specifications
Single Ringed Cytology Slide
Round
For cytology funnels, designed to collect a thin layer of cells in the ringed area
13 mm