Fisherbrand™ Round Stir Bars with Removable Pivot Ring
Stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
 View more versions of this product
Catalog No. 14-512-136
Explore available promotions
Description
Description
Stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
  • Small removable pivot ring in the center adds to their versatility
  • Pivot ring minimizes contact area of bar to vessel
  • Ring also reduces friction and the marring of plastic containers
  • No pivot ring is included on the two smallest sizes, Cat. Nos. 14-512-140 and -141
Specifications
Specifications
12.7 mm
PTFE
PTFE Coated Alnico
White
Autoclavable
76.2 mm
3 in.
0.5 in.
Stir Bar
PTFE Coated Alnico Magnet
ISO 9001:2008
Sphere
Videos
SDS
Documents
Documents
Accessories