Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Fisherbrand™ SafeSnap™ Disposable Ampule Openers
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Description
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
- To use: Slip the collar onto the ampule until collar rests on the widest part
- Grip the collar with thumb and forefinger while holding the upright ampule at bottom
- Break the ampule with a quick snap
- Dispose of the collar and ampule top together
Spécifications
Single-use | |
Ampule Opener | |
Polyethylene | |
1 to 4mL ampule |