Fisherbrand™ SafeSnap™ Disposable Ampule Openers
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
 View more versions of this product
Catalog No. 01-216-2
Explore available promotions
Description
Description
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
Polyethylene ampule collars protect fingers while breaking open the scored neck of glass ampules.
  • To use: Slip the collar onto the ampule until collar rests on the widest part
  • Grip the collar with thumb and forefinger while holding the upright ampule at bottom
  • Break the ampule with a quick snap
  • Dispose of the collar and ampule top together
Specifications
Specifications
Single-use
Ampule Opener
Polyethylene
5 to 10mL ampule