Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Lead and mercury free
Description
Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Specifications
Thermo Hydrometer | |
NIST | |
0 to 12° | |
Lead and Mercury Free |
0.1 | |
385 mm | |
Soda Lime Glass | |
For determining the percentage by weight of sucrose |