Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Lead and mercury free
 View more versions of this product
Catalog No. 11-555-213
Explore available promotions
Description
Description
Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Specifications
Specifications
Thermo Hydrometer
NIST
39 to 51°
Lead and Mercury Free
0.1
385 mm
Soda Lime Glass
For determining the percentage by weight of sucrose