Fisherbrand™ Safety Blue Thermohydrometer, Celsius
Lead and mercury free
$155.28 - $155.28
Specifications
Type | Thermo Hydrometer |
---|---|
Certifications/Compliance | NIST |
Ballast Type | Lead and Mercury Free |
For Use With (Application) | For determining the percentage by weight of sucrose |
Catalog Number | Mfr. No. | Brix Range | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Brix Range | Price | Quantity | |||||
11-555-209
|
N/A | 0 to 12° |
Each for $155.28
|
|
|||||
11-555-210
|
N/A | 9 to 21° |
Each for $155.28
|
|
|||||
11-555-212
|
N/A | 29 to 41° |
Each for $155.28
|
|
|||||
11-555-211
|
N/A | 19 to 31° |
Each for $155.28
|
|
|||||
11-555-213
|
N/A | 39 to 51° |
Each for $155.28
|
|
|||||
11-555-214
|
N/A | 49 to 61° |
Each for $155.28
|
|
|||||
11-555-216
|
N/A | 69 to 81° |
Each for $155.28
|
|
|||||
11-555-215
|
N/A | 59 to 71° |
Each for $155.28
|
|
|||||
11-555-217
|
N/A | 79 to 91° |
Each for $155.28
|
|
|||||
11-555-219
|
N/A | 35 to 70° |
Each for $155.28
|
|
|||||
11-555-208
|
N/A | -5 to 5° |
Each for $155.28
|
|
|||||
11-555-218
|
N/A | 0 to 35° |
Each for $155.28
|
|
|||||
Description
Hydrometer with enclosed thermometer to indicate temperature of the liquid under test.
Specifications
Thermo Hydrometer | |
NIST | |
Lead and Mercury Free | |
For determining the percentage by weight of sucrose |