Fisherbrand™ Screw Caps for Disposable Glass Tubes
Phenolic caps have cemented-in rubber liners
 View more versions of this product
Catalog No. 14-957-82E
Explore available promotions
Description
Description
Phenolic caps have cemented-in rubber liners
  • Autoclavable
  • Polypropylene cap is linerless
Specifications
Specifications
No
18-415
Autoclavable
Black
14-959-37A,14-957-76E, 14-959-37C
Screw Cap
Rubber
Phenolic
Non-sterile