Fisherbrand™ Serialized Class A Graduated Cylinders with Certificate of Calibration
Calibrated To Contain
 View more versions of this product
Catalog No. 08-559FC
Explore available promotions
Includes: Traceability Certification and Certificates of Calibration that list apparent volumes and calibration correction factors.
Description
Description
Calibrated To Contain.
  • White scale graduations
  • Borosilicate glass
Order Info
Case of 1
Specifications
Specifications
Cs. of 1
±2 mL
14.7 in.
White
375 mm
500 mL
White scale
Autoclavable
5mL
Round
Graduated Cylinder
To Contain
A
Glass
Traceable NIST Certificate
5 mL
±2.00 mL
Videos
SDS
Documents
Documents
Accessories