missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Silicone Recessed Septum Stoppers

Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal

$65.79 - $235.10

Specifications

Color Translucent
Material Silicone
Product Type Septum Stopper
For Use With (Application) Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
View More Specs

Products 4
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
FB57895
View Documents
N/A 9.22 to 9.75 mm
Pack of 10 Each for $65.79
 
FB57896
View Documents
N/A 13.75 to 14.25 mm
Pack of 10 Each for $106.24
 
FB57898
View Documents
N/A 20.00 to 21.00 mm
Pack of 10 Each for $235.10
 
FB57897
View Documents
N/A 14.50 to 15.50 mm
Pack of 10 Each for $102.48
 
Description

Description

  • Elliptical flange to allow efficient removal of stopper from aperature
  • Easily pierced with a hypodermic needle

Specifications

Specifications

Translucent
Silicone
Septum Stopper
Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'