Fisherbrand™ Spatula with Stainless-Steel Blade
Fisherbrand Spatula with Stainless-Steel Blade are non-stick, corrosion-free with a bright finish.
$16.04 - $32.48
Specifications
Product Type | Spatula |
---|---|
Finish | Bright |
Blade Material | Stainless Steel |
Handle Material | Hardwood |
Handle Type | Riveted |
Catalog Number | Mfr. No. | Length (Metric) Blade | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) Blade | Price | Quantity | ||||||
14-365C
|
N/A | 12.7 cm |
Each for $27.56
Pack of 12 Each for $330.72
|
|
||||||
14-365D
|
N/A | 15.24 cm |
Each for $20.41
Pack of 12 Each for $244.92
|
|
||||||
14-365E
|
N/A | 20.32 cm |
Each for $23.44
Pack of 12 Each for $281.28
|
|
||||||
14-365B
|
N/A | 10.16 cm |
Each for $16.04
Pack of 12 Each for $192.48
|
|
||||||
14-365A
|
N/A | 7.62 cm |
Each for $18.07
Pack of 12 Each for $216.84
|
|
||||||
14-365F
|
N/A | 25.4 cm |
Each for $30.37
Pack of 12 Each for $364.44
|
|
||||||
14-365G
|
N/A | 30.48 cm |
Each for $32.48
Pack of 12 Each for $389.76
|
|
||||||
Description
- Fisherbrand™ Spatula with Stainless-Steel Blade are non-stick, corrosion-free with a bright finish
- Multipurpose - Useful in educational, research or healthcare applications
- Multiple sizes allow for varied laboratory applications
- Made from ASTM stainless steel
- Non-sticking, corrosion-free standard blade
- Riveted, smooth hardwood handle designed and engineered to fit comfortably in your hand
- Electronically passivated for corrosion resistance
- Autoclavable
- Latex-free
Specifications
Spatula | |
Stainless Steel | |
Riveted |
Bright | |
Hardwood |