Fisherbrand™ Potassium Iodide Hydrogen Sulfide Test
Quickly detect Hydrogen Sulfide in water in concentrations as low as 5ppm.
Supplier:  Fisherbrand™ PRL17524V100
Catalog No. 14-862
Explore available promotions
Description
Description
  • Use this strip to test for the presence of hydrogen sulfide
Order Info
100 Strips per vial
Specifications
Specifications
Use this strip to test for the presence of hydrogen sulfide
100 Test Strips/Vial
24 Pack
Lead Acetate Test Paper