Fisherbrand™ Square Disposable Petri Dish with Grid
Ideal for phage typing, probe assays, susceptibility testing and plate counts
Catalog No. 08-757-11A
Explore available promotions
Description
Description
  • Grid consists of thirty-six 13 x 13mm squares
  • Marked numerically in one direction and alphabetically in the other
  • Disposable polystyrene
  • Radiation sterilized
Order Info
Packed 10 per polyethylene sleeve
Specifications
Specifications
Single-use
Square
Sleeve
15 mm
Polystyrene
Sterile
50 Sleeves of 10 ea.