Fisherbrand™ Standard Class B Graduated Cylinders
Calibrated To Deliver
 View more versions of this product
Catalog No. 08-550D
Explore available promotions
Description
Description
Calibrated To Deliver
  • White-ceramic graduations
  • Heavy round bases
  • Handy pouring spouts
  • Accuracy is assured even at the lowest graduations; first 10% volume is not graduated
Specifications
Specifications
Round
Graduated Cylinder
To Deliver
Round
190 mm
50 mL
White ceramic graduations
Autoclavable
1mL
±0.5 mL
7.4 in.
White
B
Borosilicate Glass
Accuracy assured even at lowest graduations
1 mL
+/- 0.50mL
Videos
SDS
Documents
Documents
Accessories