missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Stat Labels
Flourescent red labels to indicate tests are urgent.
$6.45 - $10.17
Specifications
Adhesive Type | Permanent |
---|---|
Color | Red |
Format | Label |
Product Type | Stat Label |
For Use With (Application) | Shipping |
Description
- Bright red labels with permanent adhesion
- Available in two sizes
Order Info
1000 labels per roll
Specifications
Permanent | |
Label | |
Shipping |
Red | |
Stat Label | |
1000/Roll |