Fisherbrand™ Sterile Plastic Culture Tubes: Clear Polystyrene
Ideal for tissue culture procedures.
 View more versions of this product
Catalog No. 14-956-6C
Explore available promotions
Description
Description
Ideal for tissue culture procedures
  • Gamma-irradiated and tested for sterility; ready to use
  • No chance of residual contamination
  • All screw-cap tubes have outside threads and unconstricted necks, making filling and pipetting easier
  • All snap caps can be positioned for aerobic or anaerobic applications
Order Info
Sterile caps are available separately; contact Fisher Customer Service.
Specifications
Specifications
Clear
100 mm
Ideal for tissue culture procedures
Sterile
Not Included
14 mL
Polystyrene
Bulk - 8 Pks. of 125
17 mm
Culture Tube