Fisherbrand™ Straight Connectors with Tapered Ends
Very durable.
 View more versions of this product
Catalog No. 15-315-28F
Explore available promotions
Description
Description
Very durable.
  • Excellent chemical resistance
  • Serrated connecting surfaces to prevent tubing slippage
  • Autoclavable Polypropylene
  • Adapters allow for connecting different diameter tubing
Specifications
Specifications
Tubing Connector
0.5 to 0.625 in.
2.6 in.
12.7 to 15.9mm
0.8 in.
White
Autoclavable
Polypropylene
66 mm
20.3 mm
Straight