missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Superfrost™ Disposable Microscope Slides
Superfrost coating impervious to most common laboratory chemicals and reagents
Description
Superfrost coating impervious to most common laboratory chemicals and reagents
- Brilliant white, opaque coating takes and retains most laboratory marking methods
- Each side has a written key for easy orientation; the word “SPECIMEN” instantly tells you which side is up
- Slides can be imprinted with your laboratory or hospital's name (call your Fisher HealthCare™ Sales Representative for details)
Order Info
Come in handy one-at-a-time dispenser boxes of 72 slides.
Specifications
Disposable Microscope Slide | |
1 mm | |
25 mm | |
Opaque | |
Most laboratory applications | |
1 in. |
SuperFrost | |
76 mm | |
White | |
Superfrost White Tab Slide, Full Size | |
3 in. | |
3 x 1 in. (75 x 25 mm) |