Fisherbrand™ PTFE Starburst Stirring Heads Individual Heads
Provide better mixing at bottom of vessel with less splashing.
 View more versions of this product
Catalog No. 14-513-76
Explore available promotions
Description
Description
Provide better mixing at bottom of vessel with less splashing.
  • Shape creates a deep vortex
  • Provides quiet, stable operation
  • Alnico magnet encapsulated in FDA grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable
Specifications
Specifications
19 mm
0.75 in.
White
Autoclavable
19 mm
PTFE
Stirring Head
Alnico Magnet
ISO 9001:2008
Videos
SDS
Documents
Documents
Accessories