Fisherbrand™ Traceable™ 24-Hour Stopwatches
Digital, handheld
 View more versions of this product
Catalog No. 14-649-19
Explore available promotions
Description
Description
Digital, handheld, for general-purpose applications. Fisherbrand™ Traceable 24-Hour Stopwatches provide single-action timing, time-out, continuous timing, and cumulative split functions.
  • Resolution: 1 second
  • Clock displays time of day to the second, AM/PM, date, and day of the week
  • Rugged ABS plastic case
Certifications
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology (NIST™). Calibrated by an ISO 17025 calibration laboratory accredited by A2LA™.
Specifications
Specifications
Digital Stopwatch
0.0001
LCD
0.62 in.
0.25 in.
2.12 in.
0.87 oz.
1 Year Silver Oxide battery, shockproof ABS Plastic case and lanyard
ISO 17025, A2LA, NIST
24 hr.
0.01 sec.
ABS Plastic
1.57 cm
0.63 cm
5.3 cm
0.024 kg
Ideal for lab or plant use