missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer Edge
Edge

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.

 View more versions of this product

Catalog No. 06-664-11

Edge
Explore available promotions

Description

Description

Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer includes a probe which is sealed in glycol solution and insulates sensor when refrigerator door is opened.

  • Triple display simultaneously shows Min/max and current temperatures, °F/°C switchable
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
  • 15-077-720 has a 5mL bottle designed for vaccines; 06-664-11 has a 10mL bottle
  • Glass bottle filled with glycol mimics vaccine temperature and fits in trays
  • Probe is sealed in glycol solution which insulates sensor when refrigerator door is opened
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Minimum/maximum memory monitors high and low readings overnight, on weekends, or for any time period
  • Alarm is programmable in 1° increments, sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • High-impact, chemical-resistant ABS plastic case
  • Temperature range: -50 to +70°C/-58 to +158°F
  • Resolution: 1° accuracy: ±1C
  • Battery:AA Alkaline (1)

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It provides traceability to standards provided by NIST.

Specifications

Specifications

Glycol
ISO 17025, A2LA, NIST
Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
19 mm
10 in.
25.4 cm
Refrigerator/Freezer Thermometer with 10mL bottle
4 oz.
2.75 in.
2.75 in.
70°C
-50°C
-50°C to +70°C
Refrigerators/Freezers
±1°C
70 mm
10 mL
2.75 x 2.75 x 0.75 in. (69.85 x 70 x 19 mm)
0.75 in.
Velcro™ and magnetic mounting strip, one AA battery, ABS Plastic case, alarm set in 1° increments alerts user with visual and audible signals if temperature goes out
70 mm
ABS Plastic
1°C
0.11 kg
Fridge/Freezer Digital Thermometer
158°F
-58°F
-58°F to +158°F
Yes
AA Alkaline (1)
Digital LCD
Videos
SDS
Documents

Documents

Promotions

Promotions