Fisherbrand™ Turnover Septum Stoppers
For narrow-aperture containers
$49.76 - $151.59
Specifications
Product Type Turnover Septum Stopper
For Use With (Application) For narrow aperture containers
Total Positions 1
View More Specs
Products 26
Catalog Number Mfr. No. Color Diameter (Metric) Inner Size For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Color Diameter (Metric) Inner Size For Use With (Equipment) Price Quantity  
FB57872
View Documents
N/A Red 6.5 mm 9 mm For 0.77 to 0.82cm I.D. containers
Pack of 10 Each for $49.76
FB57873
View Documents
N/A Red 8 mm 13 mm For 0.92 to 0.97cm I.D. containers
Pack of 10 Each for $54.66
FB57874
View Documents
N/A Red 9.5 mm 17 mm For 1.1 to 1.15cm I.D. containers
Pack of 10 Each for $51.31
FB57875
View Documents
N/A Red 11 mm 21 mm For 1.2 to 1.3cm I.D. containers
Pack of 10 Each for $56.79
FB57878
View Documents
N/A Red 16 mm 33 mm For 1.7 to 1.75cm I.D. containers
Pack of 10 Each for $52.38
FB57877
View Documents
N/A Red 14 mm 29 mm For 1.5 to 1.6cm I.D. containers
Pack of 10 Each for $52.38
FB57876
View Documents
N/A Red 12.5 mm 25 mm For 1.3 to 1.35cm I.D. containers
Pack of 10 Each for $51.00
FB57880
View Documents
N/A Red 19 mm 41 mm For 2 to 2.1cm I.D. containers
Pack of 10 Each for $95.00
FB57881
View Documents
N/A Red 20 mm 45 mm For 2.2 to 2.3cm I.D. containers
Pack of 10 Each for $88.79
FB57884
View Documents
N/A Red 25 mm 57 mm For 2.65 to 2.75cm I.D. containers
Pack of 10 Each for $145.55
FB57883
View Documents
N/A Red 24 mm 53 mm For 2.55 to 2.65cm I.D. containers
Pack of 10 Each for $151.59
FB68682
View Documents
N/A Cream 9.5 mm 17 mm For 1.1 to 1.15cm I.D. containers
Pack of 10 Each for $51.31
FB68681
View Documents
N/A Cream 8 mm 13 mm For 0.92 to 0.97cm I.D. containers
Pack of 10 Each for $54.66
FB68683
View Documents
N/A Cream 11 mm 21 mm For 1.1 to 1.15cm I.D. containers
Pack of 10 Each for $56.79
FB68685
View Documents
N/A Cream 14 mm 29 mm For 1.5 to 1.6cm I.D. containers
Pack of 10 Each for $52.38
FB68684
View Documents
N/A Cream 12.5 mm 25 mm For 1.3 to 1.35cm I.D. containers
Pack of 10 Each for $52.38
FB68686
View Documents
N/A Cream 16 mm 33 mm For 1.7 to 1.75cm I.D. containers
Pack of 10 Each for $64.31
FB68687
View Documents
N/A Cream 17.5 mm 37 mm For 1.77 to 1.87cm I.D. containers
Pack of 10 Each for $75.79
FB68688
View Documents
N/A Cream 19 mm 41 mm For 2 to 2.1cm I.D. containers
Pack of 10 Each for $95.00
FB68689
View Documents
N/A Cream 20 mm 45 mm For 2.2 to 2.3cm I.D. containers
Pack of 10 Each for $92.07
FB68690
View Documents
N/A Cream 22 mm 49 mm For 2.35 to 2.45cm I.D. containers
Pack of 10 Each for $116.45
FB68691
View Documents
N/A Cream 24 mm 53 mm For 2.55 to 2.65cm I.D. containers
Pack of 10 Each for $151.52
FB68692
View Documents
N/A Cream 25 mm 57 mm For 2.65 to 2.75cm I.D. containers
Pack of 10 Each for $147.41
FB57879
View Documents
N/A Red 17.5 mm 37mm For 1.77 to 1.87cm I.D. containers
Pack of 10 Each for $75.86
FB57882
View Documents
N/A Red 6.5 mm 49mm For 0.77 to 0.82cm I.D. containers
Pack of 10 Each for $116.45
FB68680
View Documents
N/A Cream 6.5 mm 9mm For 0.77 to 0.82cm I.D. containers
Pack of 10 Each for $49.76
Description
Description
For narrow-aperture containers
  • External serrations on plug
  • Turnover sleeve folds down over the container's outer neck and creates a secure seal
  • Easily perforated with hypodermic needles
  • Available in red or cream rubber in packs of 10
Specifications
Specifications
Turnover Septum Stopper
For narrow aperture containers
1
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'