Fisherbrand™ Three-hole Twistit™ Rubber Stoppers
Stoppers made from soft, resilient, natural rubber with safe convenient countersunk holes, stoppers feature excellent aging characteristics with high resistance to deformation
Description
- Simple twist-off seals provide convenient one, two, or three-hole use
- Solid stoppers are vacuum-tested at 30 in. of mercury
- High resistance to deformation—even after repeated autoclaving
- Each stopper has its size number in raised figures on top
Specifications
Black | |
6 | |
6 | |
3 |
Stopper (3 holes) | |
Multi-purpose applications | |
Natural Rubber |