Fisherbrand™ Ultra Flex Clamping Systems
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
Supplier:  Fisherbrand™ 80862754
 View more versions of this product
Catalog No. 02-217-009
Explore available promotions
Description
Description
Unique flexible arms that are extremely versatile and can be placed in virtually any position or angle.
  • Ultra Flex comes in three different systems: base plate, lab-frame connector, and bench clamp
  • Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head, nickel-plated flex arm in 305mm (12 in.) or 457mm (18 in.) length, and utility wrench
  • 2-prong and 3-prong clamp heads are also supplied with non-slip vinyl sleeves and, for temperatures above 100°C, fiberglass covers
Specifications
Specifications
1 Ea.
Fiberglass (Cover), Vinyl (Sleeve)
Ultra flex clamping system components
13 in.
7.49, 6.9, 1.29 cm
2-Prong, 3-Prong
Ultra Flex Clamp System
Lab frame connector
Ultra Flex 12 with Lab-Frame Connector
33.02 cm
2.95, 2.72, 0.51 in.