Fisherbrand™ Test Tube Holder for Ultrasonic Units
Use this Fisherbrand™ Test Tube Holder with Fisherbrand™ 112xx Series Dual Frequency Ultrasonic Cleaners. Easily support and handle test tubes within the ultrasonic cleaner.
Supplier:  Fisherbrand™ 2070770000
Catalog No. 29-406-67
Explore available promotions
Description
Description
  • Multiple holes to support 12 test tubes
  • Dual handles easily secures holder in the ultrasonic cleaner
  • Stainless steel
  • Light and easy to maneuver
Specifications
Specifications
Test Tube Holder
FB11211 Ultrasonic Cleaning Unit
Stainless Steel