Fisherbrand™ Universal Pipet Pump Edge
Edge
Pipet filler with easy one-hand operation provides accurate titration. Color-coded for easy identification.
 View more versions of this product
Catalog No. 14-955-110
Edge
Explore available promotions
Description
Description
  • Made from high-grade polyethylene
  • Accepts glass and serological pipets
  • Ideal for soap and acid titrations
  • Clear markings make measuring easy
  • 10mL pipet marked in 0.10mL increments
  • 25mL pipet marked in 0.20mL increments
  • Rotating thumbwheel draws or dispensed liquid
  • Side lever allows entire contents to be rapidly dispensed
25mL fits both 10mL and 25mL pipets
Specifications
Specifications
Green
Polyethylene
Pipet Controller
10 mL