Oak Ridge Urine Sediment Tubes
Designed for use with Sysmex™ and Siemens™ Atlas™ instruments
Supplier: Oak Ridge 02014100
Description
- Non-flared top for efficient nesting in tube racking systems
- Circular in shape
- Constructed of shatter-resistant polystyrene
- Transparent body allows for easy viewing of the specimen
Specifications
Urine Sediment Tube | |
Polystyrene | |
1000/Cs. |