Fisherbrand™ Urine Sedimentation System: Bulb Decanting Pipette
The Fisherbrand™ Urine Sedimentation System is designed to produce the same results as the most popular style urine sedimentation system using the same test procedure
Catalog No. 14-375-560
Explore available promotions
Description
Description
Decanting pipet allows supernatant to be decanted off; leaves 1mL of urine-containing sediment in the bottom of the tube.
Specifications
Specifications
Bulb Decanting Pipette
Designed to produce the same results as the most popular style Urine sedimentation system using the same test procedure
500 Case