Fisherbrand™ UriSystem™ DeciSlide™ 10-Test Slide
Analyze up to 10 samples simultaneously
Description
- Made of optically clear acrylic
- Features an integral cover slip 10 individual chambers
- Each chamber is sealed on three sides to prevent cross contamination
- Reduces cross-contamination while standardizing volume per area
Specifications
Urine Sedimentation Test Slide | |
Acrylic | |
Clear | |
Urine sedimentation analysis | |
1000/Cs. |
Integral Coverslip | |
Flat | |
Non-Gridded | |
Non-Gridded | |
100 Pk. |