Fisherbrand™ Verification Thermometers for Ovens, Incubators, Freezers, and Refrigerators
Insertion of the thermometer into the included recyclable plastic bottle protects samples from contamination by eliminating the need for direct contact.
 View more versions of this product
Catalog No. 13201601
Explore available promotions
Description
Description
  • Mercury-free liquid-in-glass thermometers with glass bead bottle fill
  • Bottle fill mimics the samples being stored for more accurate temperature monitoring
  • Features permanently fused markings and lead free, white back glass
  • Individually serialized; traceable to NIST
Ideal for chemical industry, petrochemistry, pharmaceutical, medical, cosmetic, producers of chemicals, university, dairies, breweries, distillers, beer cellars, refrigeration house, hot houses, scientific institutes, blood banks, refrigeration/freezer/incubator/oven equipment producers and producers of laboratory equipment.
Specifications
Specifications
NIST
60°F
20°F to 60°F
2°F
Refrigerator
20°F