Fisherbrand™ Verification Thermometers for Ovens, Incubators, Freezers, and Refrigerators
Insertion of the thermometer into the included recyclable plastic bottle protects samples from contamination by eliminating the need for direct contact.
Supplier:  Fisherbrand™ B606001320
 View more versions of this product
Catalog No. 13201602
Explore available promotions
Description
Description
  • Mercury-free liquid-in-glass thermometers with glass bead bottle fill
  • Bottle fill mimics the samples being stored for more accurate temperature monitoring
  • Features permanently fused markings and lead free, white back glass
  • Individually serialized; traceable to NIST
Ideal for chemical industry, petrochemistry, pharmaceutical, medical, cosmetic, producers of chemicals, university, dairies, breweries, distillers, beer cellars, refrigeration house, hot houses, scientific institutes, blood banks, refrigeration/freezer/incubator/oven equipment producers and producers of laboratory equipment.
Specifications
Specifications
Glass beads
Oven
50°C
50°C to 200°C
NIST
200°C
2.5°C