Fisherbrand™ Write-On Tape
Sticks to glass, plastic, paper and wood.
 View more versions of this product
Catalog No. 15948
Explore available promotions
Description
Description
  • Can be used in temperatures from -23°C to +121°C (-9°F to +250°F)
  • Well suited for use in autoclaves, refrigerators, deep freeze, sterilizers, and incubators
  • Easily marked with pencil, pen, ballpoint and most felt-tip markers
  • Removes without residue
  • Resists oil, water, and acid
Specifications
Specifications
White
Write-On™ Tape
1 Ea.
-23°C to +121°C
1.5 in.
Tape
Autoclave, Refrigerator/freezer, Sterilizer, Incubator
-9 to 250°F
14 yd.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories