Fisherbrand™ Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
 View more versions of this product
Catalog No. 01-816F
Explore available promotions
Description
Description
Zipper-locking feature securely seals bags
  • Durable, transparent polyethylene
  • Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
  • Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Specifications
Not Autoclavable
Zip Seal
Polyethylene
50
26 x 9 in. (66 x 23cm)
66 cm
23 cm
5 lb.
Clear
200
4 mil
26 in.
9 in.