Fisherbrand™ Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
 View more versions of this product
Catalog No. 01-816C
Explore available promotions
Description
Description
Zipper-locking feature securely seals bags
  • Durable, transparent polyethylene
  • Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
  • Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Specifications
Not Autoclavable
Zip Seal
Polyethylene
50
18 x 9 in. (46 x 23cm)
46 cm
23 cm
3 lb.
Clear
250
4 mil
18 in.
9 in.